CAS RN 412940-35-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Mitochondrial Citrate Transport Protein (CTP) Inhibitor
The Mitochondrial Citrate Transport Protein (CTP) Inhibitor, also referenced under CAS 412940-35-3, controls the biological activity of Mitochondrial Citrate Transport Protein (CTP).