Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Mitochondrial Citrate Transport Protein (CTP) Inhibitor

Catalog No. 4758775MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-587-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-587-75MG Supplier MilliporeSigma™ Supplier No. 4758775MG
Add to Cart
Add to Cart

The Mitochondrial Citrate Transport Protein (CTP) Inhibitor, also referenced under CAS 412940-35-3, controls the biological activity of Mitochondrial Citrate Transport Protein (CTP).

The Mitochondrial Citrate Transport Protein (CTP) Inhibitor, also referenced under CAS 412940-35-3, controls the biological activity of Mitochondrial Citrate Transport Protein (CTP).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
CAS 412940-35-3
Molecular Formula C13H9ClN2O6S
Formula Weight 356.7g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.