CAS RN 41317-15-1
IUPAC Name:
4-methoxy-2-methyl-N-phenylaniline
Synonyms:
4-methoxy-2-methyl-N-phenylaniline
Molecular Weight (g/mol):
213.28
Molecular Formula:
C14H15NO
InChi Key:
CYMPUOGZUXAIMY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(NC2=CC=CC=C2)C(C)=C1
1
–
1
of
1
results
1
4-Methoxy-2-methyldiphenylamine 97.0+%, TCI America™
CAS: 41317-15-1 Molecular Formula: C14H15NO Molecular Weight (g/mol): 213.28 MDL Number: MFCD00272619 InChI Key: CYMPUOGZUXAIMY-UHFFFAOYSA-N PubChem CID: 162461 IUPAC Name: 4-methoxy-2-methyl-N-phenylaniline SMILES: CC1=C(C=CC(=C1)OC)NC2=CC=CC=C2