Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2-methyldiphenylamine 97.0+%, TCI America™
SDP

Catalog No. M128625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M128625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M128625G Supplier TCI America Supplier No. M128625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41317-15-1
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD00272619
InChI Key CYMPUOGZUXAIMY-UHFFFAOYSA-N
PubChem CID 162461
IUPAC Name 4-methoxy-2-methyl-N-phenylaniline
SMILES CC1=C(C=CC(=C1)OC)NC2=CC=CC=C2

Specifications

Melting Point 81°C
Boiling Point 171°C
Quantity 25 g
Formula Weight 213.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-methyldiphenylamine
EINECSNumber (3)-3869
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.