accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OHONH

CAS RN 41340-25-4

IUPAC Name: 2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
Synonyms: etodolac etodolac, (-)-isomer ultradol etodolac (product) ramodar monosodium salt etodolac etodolac, (-)- etodolac (substance) etodolac, (+-)- etodolac, (S)-
Molecular Weight (g/mol): 287.36
Molecular Formula: C17H21NO3
InChi Key: NNYBQONXHNTVIJ-UHFFFAOYNA-N
SMILES: CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results