accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOHO

CAS RN 41345-70-4

IUPAC Name: 3-(acetylsulfanyl)propanoic acid
Synonyms: 3-(acetylsulfanyl)propanoic acid
Molecular Weight (g/mol): 148.18
Molecular Formula: C5H8O3S
InChi Key: AYQXANXXZYKTDL-UHFFFAOYSA-N
SMILES: CC(=O)SCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results