accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41372-22-9

IUPAC Name: calcium bis(2-hydroxypropanoate) hydrate
Synonyms: calcium hydrate dilactate
Molecular Weight (g/mol): 236.23
Molecular Formula: C6H12CaO7
InChi Key: MDAVASCOAJMZHZ-UHFFFAOYNA-L
SMILES: O.[Ca++].CC(O)C([O-])=O.CC(O)C([O-])=O
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results