Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-™-Propargyl-Ala-Oh

Catalog No. F53121G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F53121G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. F53121G Supplier Sigma Aldrich Supplier No. F53121G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alkyne amino acid, more stable for peptide stapling via click chemistry than propargylglycine.

Specifications

CAS 41372-22-9
Molecular Formula C6H10CaO6 -+ xH2O
Linear Formula [CH3CH(OH)COO]2Ca -+ xH2O
MDL Number MFCD00150344
Quantity 250 g
Molecular Weight (g/mol) 218.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.