CAS RN 41375-88-6
IUPAC Name:
3-(16-methylheptadecyl)oxolane-2,5-dione
Synonyms:
3-(16-methylheptadecyl)oxolane-2,5-dione
Molecular Weight (g/mol):
352.56
Molecular Formula:
C22H40O3
InChi Key:
XRAJHWOOOMPDFP-UHFFFAOYNA-N
SMILES:
CC(C)CCCCCCCCCCCCCCCC1CC(=O)OC1=O
1
–
1
of
1
results
1
Isooctadecylsuccinic Anhydride (mixture of branched chain isomers) 90.0+%, TCI America™
CAS: 41375-88-6 Molecular Formula: C22H40O3 Molecular Weight (g/mol): 352.56 MDL Number: MFCD00047132 InChI Key: XRAJHWOOOMPDFP-UHFFFAOYNA-N Synonym: iso-Octadecylsuccinic Anhydride PubChem CID: 536991 IUPAC Name: 3-(16-methylheptadecyl)oxolane-2,5-dione SMILES: CC(C)CCCCCCCCCCCCCCCC1CC(=O)OC1=O