Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isooctadecylsuccinic Anhydride (mixture of branched chain isomers) 90.0+%, TCI America™
SDP

Catalog No. I014625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I014625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I014625G Supplier TCI America Supplier No. I014625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41375-88-6
Molecular Formula C22H40O3
Molecular Weight (g/mol) 352.56
MDL Number MFCD00047132
InChI Key XRAJHWOOOMPDFP-UHFFFAOYNA-N
Synonym iso-Octadecylsuccinic Anhydride
PubChem CID 536991
IUPAC Name 3-(16-methylheptadecyl)oxolane-2,5-dione
SMILES CC(C)CCCCCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Formula Weight 352.56
Percent Purity ≥90.0% (T)
Chemical Name or Material Isooctadecylsuccinic Anhydride (mixture of branched chain isomers)
EINECSNumber (2)-0851
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.