accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 41424-11-7

IUPAC Name: 4'-(hexyloxy)-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-(hexyloxy)-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 279.38
Molecular Formula: C19H21NO
InChi Key: YUYXUPYNSOFWFV-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results