Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4'-(Hexyloxy)-4-biphenylcarbonitrile

Catalog No. 3386561G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-865-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 33-865-61G Supplier Sigma Aldrich Supplier No. 3386561G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4'-(Hexyloxy)-4-biphenylcarbonitrile

Specifications

CAS 41424-11-7
Melting Point 56°C to 58°C
Molecular Formula C19H21NO
Linear Formula CH3(CH2)5 OC6H4C6H4CN
Quantity 1 g
MDL Number MFCD00134145
Synonym 4 -(Hexyloxy)-4-biphenyl-carbonitrile; 4 -(Hexyloxy)[1,1 -biphenyl]-4-carbonitrile; 4 -Hexyloxy-4-cyanobiphenyl; 4 -Hexyloxybiphenylyl-4-carbonitrile; 4-(4-Hexyloxyphenyl)benzonitrile; 4-(n-Hexyloxy)-4 -cyanobiphenyl; 4-Cyano-4 -(hexyloxy)biphenyl; 4-Cyano-4 -n -hexyloxybiphenyl; 4-Hexyloxy-4 -cyanobiphenyl; p -Hexyloxy-p -cyanobiphenyl
Molecular Weight (g/mol) 279.38
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.