accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONSNOONH2CH3

CAS RN 41450-33-3

IUPAC Name: ethyl 2-(2-amino-5-nitrophenyl)-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-(2-amino-5-nitrophenyl)-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 307.32
Molecular Formula: C13H13N3O4S
InChi Key: ASHVHIIHZOANMU-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=C(N)C=CC(=C1)[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results