accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNOONH2

CAS RN 41450-34-4

IUPAC Name: 4-nitro-2-(4-phenyl-1,3-thiazol-2-yl)aniline
Synonyms: 4-nitro-2-(4-phenyl-1,3-thiazol-2-yl)aniline
Molecular Weight (g/mol): 297.33
Molecular Formula: C15H11N3O2S
InChi Key: FSEYQHXDVSVHJO-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1)[N+]([O-])=O)C1=NC(=CS1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results