accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHNOONHNH2

CAS RN 4146-43-4

IUPAC Name: butanedihydrazide
Synonyms: succinic dihydrazide
Molecular Weight (g/mol): 146.15
Molecular Formula: C4H10N4O2
InChi Key: HCOMFAYPHBFMKU-UHFFFAOYSA-N
SMILES: NNC(=O)CCC(=O)NN
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)