Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinic Dihydrazide 97.0+%, TCI America™
SDP

Catalog No. S048225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S048225G 25 g
S0482100G 100 g
S0482500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S048225G Supplier TCI America Supplier No. S048225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4146-43-4
Molecular Formula C4H10N4O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00007613
InChI Key HCOMFAYPHBFMKU-UHFFFAOYSA-N
Synonym Butanediohydrazide
PubChem CID 72491
IUPAC Name butanedihydrazide
SMILES NNC(=O)CCC(=O)NN

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 25 g
Formula Weight 146.15
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Succinic Dihydrazide
EINECSNumber (2)-0902
RTECSNumber WM7500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.