CAS RN 4151-50-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PESTANAL™ Sulfluramid, ≥98% (GC), MilliporeSigma™ Supelco™
Sulfluramid is a slow-acting insecticide for controlling ants.