Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Sulfluramid, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018190 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8190 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-8190 Supplier MilliporeSigma Supelco Supplier No. 9124225MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sulfluramid is a slow-acting insecticide for controlling ants.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4151-50-2
Product Line PESTANAL
Physical Form Neat
Linear Formula C10H6F17 NO2 S
MDL Number MFCD00042274
Synonym N-Ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide; N-Ethylperfluorooctane-1-sulfonamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 527.20
Percent Purity ≥98% (GC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Carbon content, 22.3-23.3 wt.%
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.