accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHCl

CAS RN 4151-97-7

IUPAC Name: 1-chloro-3-methoxypropan-2-ol
Synonyms: 1-chloro-3-methoxypropan-2-ol
Molecular Weight (g/mol): 124.56
Molecular Formula: C4H9ClO2
InChi Key: FOLYKNXDLNPWGC-UHFFFAOYNA-N
SMILES: COCC(O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results