Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-methoxy-2-propanol 98.0+%, TCI America™
SDP

Catalog No. C147225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C147225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C147225G Supplier TCI America Supplier No. C147225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4151-97-7
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.564
MDL Number MFCD00018935
InChI Key FOLYKNXDLNPWGC-UHFFFAOYSA-N
Synonym 1-chloro-3-methoxy-2-propanol, 2-propanol, 1-chloro-3-methoxy, 3-chloro-1-methoxy-2-propanol, acmc-209jkq, folyknxdlnpwgc-uhfffaoysa, 2-propanol,1-chloro-3-methoxy, 2-hydroxy-3-methoxypropyl chloride, 3-chloro-2-hydroxypropyl methyl ether
PubChem CID 97988
IUPAC Name 1-chloro-3-methoxypropan-2-ol
SMILES COCC(CCl)O

Specifications

Color Yellow
Boiling Point 95°C
Quantity 25 g
Formula Weight 124.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-methoxy-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.