accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO2H2H2H2H2H

CAS RN 4165-60-0

IUPAC Name: 1-nitro(²H₅)benzene
Synonyms: 1-nitro(²H₅)benzene
Molecular Weight (g/mol): 128.14
Molecular Formula: C6H5NO2
InChi Key: LQNUZADURLCDLV-RALIUCGRSA-N
SMILES: [2H]C1=C([2H])C([2H])=C(C([2H])=C1[2H])[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)