accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COOCH3

CAS RN 4166-20-5

IUPAC Name: 2,5-dimethyl-4-oxo-4,5-dihydrofuran-3-yl acetate
Synonyms: 2,5-dimethyl-4-oxo-5H-furan-3-yl acetate
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: VPKIUOQJQJVLRW-UHFFFAOYNA-N
SMILES: CC1OC(C)=C(OC(C)=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results