Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetoxy-2,5-dimethyl-3(2H)-furanone 97.0+%, TCI America™
SDP

Catalog No. A219225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A219225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A219225G Supplier TCI America Supplier No. A219225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4166-20-5
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00799462
InChI Key VPKIUOQJQJVLRW-UHFFFAOYSA-N
Synonym furaneol acetate, 2,5-dimethyl-4-oxo-4,5-dihydrofuran-3-yl acetate, 4-acetoxy-2,5-dimethyl-3 2h-furanone, 4-acetoxy-2,5-dimethyl-3 2h furanone, 3 2h-furanone, 4-acetyloxy-2,5-dimethyl, 4-acetoxy-2.5-dimethyl-3 2h-furanone, 4-acetyloxy-2,5-dimethyl-3 2h-furanone, 4-hydroxy-2,5-dimethyl-3 2h-furanone acetate, 3 2h-furanone, 4-hydroxy-2,5-dimethyl-, acetate, 2,5-dimethyl-4-oxofuran-3-yl acetate
PubChem CID 61334
IUPAC Name (2,5-dimethyl-4-oxofuran-3-yl) acetate
SMILES CC1C(=O)C(=C(O1)C)OC(=O)C

Specifications

Color Yellow
Boiling Point 243°C
Quantity 25 g
Formula Weight 170.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Acetoxy-2,5-dimethyl-3(2H)-furanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.