accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOONOO

CAS RN 41663-84-7

IUPAC Name: 2-methyl-5-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-methyl-5-nitroisoindole-1,3-dione
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H6N2O4
InChi Key: JBCHWGTZAAZJKG-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=CC=C(C=C2C1=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results