Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-4-nitrophthalimide 99.0+%, TCI America™
SDP

Catalog No. M13105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13105G 5 g
M131025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13105G Supplier TCI America Supplier No. M13105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41663-84-7
Molecular Formula C9H6N2O4
Molecular Weight (g/mol) 206.157
MDL Number MFCD00454263
InChI Key JBCHWGTZAAZJKG-UHFFFAOYSA-N
PubChem CID 92321
IUPAC Name 2-methyl-5-nitroisoindole-1,3-dione
SMILES CN1C(=O)C2=C(C1=O)C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 178°C
Color Yellow
Quantity 5 g
Formula Weight 206.16
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-4-nitrophthalimide
RTECSNumber NR3398541
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.