accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONClCl

CAS RN 41667-95-2

IUPAC Name: 5,6-dichloropyridine-3-carboxylic acid
Synonyms: 5,6-dichloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: RNRLTTNKVLFZJS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(Cl)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results