Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dichloronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D24015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24015G 5 g
D240125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24015G Supplier TCI America Supplier No. D24015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 41667-95-2 | C6H3Cl2NO2 | 191.995 g/mol

Chemical Identifiers

CAS 41667-95-2
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00075181
InChI Key RNRLTTNKVLFZJS-UHFFFAOYSA-N
Synonym 5,6-dichloronicotinic acid, 5,6-dichloro nicotinic acid, 5,6-dichloro-3-pyridinecarboxylic acid, 5,6-dichloronicotinicacid, 2,3-dichloro-5-carboxypyridine, 3-pyridinecarboxylic acid, 5,6-dichloro, 5,6-dichloro-nicotinic acid, pubchem12968, acmc-209jl4, 5,6 dichloronicotinic acid
PubChem CID 247987
IUPAC Name 5,6-dichloropyridine-3-carboxylic acid
SMILES C1=C(C=NC(=C1Cl)Cl)C(=O)O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 5 g
Formula Weight 192.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichloronicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.