accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PCH3CH3

CAS RN 4168-73-4

IUPAC Name: trihexylphosphane
Synonyms: trihexylphosphane
Molecular Weight (g/mol): 286.48
Molecular Formula: C18H39P
InChi Key: FPZZZGJWXOHLDJ-UHFFFAOYSA-N
SMILES: CCCCCCP(CCCCCC)CCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results