Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trihexylphosphine 90.0+%, TCI America™
SDP

Catalog No. T100525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T100525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T100525ML Supplier TCI America Supplier No. T100525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4168-73-4
Molecular Formula C18H39P
Molecular Weight (g/mol) 286.484
MDL Number MFCD00015285
InChI Key FPZZZGJWXOHLDJ-UHFFFAOYSA-N
PubChem CID 77831
IUPAC Name trihexylphosphane
SMILES CCCCCCP(CCCCCC)CCCCCC

Specifications

Color Colorless
Boiling Point 227°C
Quantity 25 mL
Formula Weight 286.48
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Trihexylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.