accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41717-26-4

IUPAC Name: 1-[(2,4,6-trimethylphenyl)methyl]piperazine
Synonyms: 1-[(2,4,6-trimethylphenyl)methyl]piperazine
Molecular Weight (g/mol): 218.34
Molecular Formula: C14H22N2
InChi Key: FOERNUXLFALRDN-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CN2CCNCC2)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results