accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOO

CAS RN 4176-55-0

IUPAC Name: 1,3,2λ⁴-dioxathian-2-one
Synonyms: 1,3,2λ⁴-dioxathian-2-one
Molecular Weight (g/mol): 122.14
Molecular Formula: C3H6O3S
InChi Key: LOURZMYQPMDBSR-UHFFFAOYSA-N
SMILES: O=S1OCCCO1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)