Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Propylene sulfite

Catalog No. 77424325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-424-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 77-424-325G Supplier Sigma Aldrich Supplier No. 77424325G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Trimethylene sulfite is used as high temperature additive for electrolytes in Lithium ion batteries. It improves the decomposition resistance of the electrolyte.

Specifications

CAS 4176-55-0
Density 1.347 g/mL (at 25°C)
Molecular Formula C3H6O3S
Refractive Index n20/D 1.453
Linear Formula C3H6O3S
Quantity 25 g
MDL Number MFCD01682880
Synonym 2-Oxo-1,3,2-dioxathiane; PS; TMS; Trimethylene sulfite
Molecular Weight (g/mol) 122.14
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.