accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41764-74-3

H3COOCH3OHNNH2

CAS RN 41764-74-3

IUPAC Name: 3,4-dimethoxybenzohydrazide
Synonyms: 3,4-dimethoxybenzohydrazide
Molecular Weight (g/mol): 196.21
Molecular Formula: C9H12N2O3
InChi Key: LJMQIGMMUZLDOC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results