accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2HCl

CAS RN 41784-08-1

IUPAC Name: 1-(4-aminophenyl)ethan-1-one hydrochloride
Synonyms: 4-aminoacetophenone hydrochloride
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: LCYIZBVMHJGJGP-UHFFFAOYSA-N
SMILES: Cl.CC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results