Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Aminoacetophenone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A025225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A025225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A025225G Supplier TCI America Supplier No. A025225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41784-08-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00060232
InChI Key LCYIZBVMHJGJGP-UHFFFAOYSA-N
PubChem CID 23333831
IUPAC Name 1-(4-aminophenyl)ethan-1-one hydrochloride
SMILES Cl.CC(=O)C1=CC=C(N)C=C1

Specifications

Quantity 25 g
Formula Weight 171.62
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Aminoacetophenone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.