accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4181-20-8

IUPAC Name: 4-iodo-N,N-bis(4-iodophenyl)aniline
Synonyms: 4-iodo-N,N-bis(4-iodophenyl)aniline
Molecular Weight (g/mol): 623.01
Molecular Formula: C18H12I3N
InChi Key: AQGZDWJFOYXGAA-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)N(C1=CC=C(I)C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results