Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-iodophenyl)amine 97.0+%, TCI America™
SDP

Catalog No. T17245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17245G 5 g
T172425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17245G Supplier TCI America Supplier No. T17245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4181-20-8
Molecular Formula C18H12I3N
Molecular Weight (g/mol) 623.014
MDL Number MFCD01321198
InChI Key AQGZDWJFOYXGAA-UHFFFAOYSA-N
Synonym tris 4-iodophenyl amine, tris-4-iodophenyl amine, trisiodophenylamine, 4-iodo-n,n-bis 4-iodophenyl aniline, tris-4-iodo-phenyl-amine, pubchem19658, tris p-iodophenyl amine, acmc-209jlv, ksc235o2h
PubChem CID 9809282
IUPAC Name 4-iodo-N,N-bis(4-iodophenyl)aniline
SMILES C1=CC(=CC=C1N(C2=CC=C(C=C2)I)C3=CC=C(C=C3)I)I

Specifications

Melting Point 189°C
Color Yellow
Quantity 5 g
Formula Weight 623.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-iodophenyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.