accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNNH3C

CAS RN 4184-79-6

IUPAC Name: 5,6-dimethyl-1H-1,2,3-benzotriazole
Synonyms: 5,6-dimethyl-1H-1,2,3-benzotriazole|5,6-Dimethylbenzotriazole
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: MVPKIPGHRNIOPT-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2N=NNC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results