Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dimethyl-1,2,3-benzotriazole Hydrate 99.0+%, TCI America™
SDP

Catalog No. D21775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21775G Supplier TCI America Supplier No. D21775G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4184-79-6
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00149344
InChI Key MVPKIPGHRNIOPT-UHFFFAOYSA-N
Synonym 5,6-dimethyl-1h-benzotriazole hydrate, 5,6-dimethyl-1,2,3-benzotriazole hydrate, 5,6-dimethyl-1h-1,2,3-benzotriazole hydrate, acmc-1aqbb, 1h-benzotriazole,5,6-dimethyl, 5,6-dimethylbenzotriazole, oxamethane, 5,6-dimethyl-2h-benzotriazole hydrate, 5,6-dimethyl-1h-benzotriazole monohydrate, 5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
PubChem CID 16219214
IUPAC Name 5,6-dimethyl-1H-1,2,3-benzotriazole
SMILES CC1=C(C)C=C2N=NNC2=C1

Specifications

Melting Point 154°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethyl-1,2,3-benzotriazole Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.