accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41873-74-9

CAS RN 41873-74-9

IUPAC Name: 4-methylphenyl but-2-enoate
Synonyms: 4-methylphenyl but-2-enoate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: FGJQFRNEXKYBQC-UHFFFAOYSA-N
SMILES: CC=CC(=O)OC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)