Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Tolyl Crotonate 55.0+%, TCI America™
SDP

Catalog No. C0866500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0866500ML 500 mL
C086625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0866500ML Supplier TCI America Supplier No. C0866500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 41873-74-9
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00059385
InChI Key FGJQFRNEXKYBQC-UHFFFAOYSA-N
Synonym p-Cresyl Crotonate, Crotonic Acid p-Tolyl Ester, Crotonic Acid p-Cresyl Ester
PubChem CID 5352024
IUPAC Name 4-methylphenyl but-2-enoate
SMILES CC=CC(=O)OC1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 158°C
Quantity 500 mL
Formula Weight 176.22
Percent Purity ≥55.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Crotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.