accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOOCH3

CAS RN 418777-17-0

IUPAC Name: N-(4-ethylphenyl)-2-methoxybenzamide
Synonyms: N-(4-ethylphenyl)-2-methoxybenzamide
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: WLRSWQTWFMCYRS-UHFFFAOYSA-N
SMILES: CCC1=CC=C(NC(=O)C2=CC=CC=C2OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results