accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 41879-39-4

IUPAC Name: O-(tert-butyldimethylsilyl)hydroxylamine
Synonyms: O-(tert-butyldimethylsilyl)hydroxylamine
Molecular Weight (g/mol): 147.29
Molecular Formula: C6H17NOSi
InChi Key: SSUCKKNRCOFUPT-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)ON
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)