Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O-(tert-Butyldimethylsilyl)hydroxylamine

Catalog No. 3674785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-747-85G 5 g
36-747-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-747-85G Supplier Sigma Aldrich Supplier No. 3674785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

O-(tert-Butyldimethylsilyl)hydroxylamine

Specifications

CAS 41879-39-4
Melting Point 62°C to 65°C (lit.)
Boiling Point 87°C to 90°C (40 mmHg)
Molecular Formula C6H17NOSi
Linear Formula (CH3)3 CSi(CH3)2 ONH2
MDL Number MFCD00054944
Quantity 5 g
Synonym Aminoxy-tert-butyldimethylsilane
Molecular Weight (g/mol) 147.29
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.