accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3C(E/Z)

CAS RN 4188-88-9

IUPAC Name: methyl 4-oxopent-2-enoate
Synonyms: methyl 4-oxopent-2-enoate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: GLVNZYODMKSEPS-UHFFFAOYSA-N
SMILES: COC(=O)C=CC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results