Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Acetylacrylate 98.0+%, TCI America™
SDP

Catalog No. A00711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A00711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A00711G Supplier TCI America Supplier No. A00711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4188-88-9
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00059331
InChI Key GLVNZYODMKSEPS-UHFFFAOYSA-N
Synonym Acetylacrylic Acid Methyl Ester

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 90°C
Quantity 1 g
Formula Weight 128.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Acetylacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.