accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(E)

CAS RN 4189-02-0

IUPAC Name: (2E)-undec-2-enoic acid
Synonyms: trans-2-undecenoic acid (2E)-2-undecenoic acid
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: IGBBVTAVILYDIO-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results