Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Undecenoic Acid 70.0+%, TCI America™
SDP

Catalog No. U00325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00325G Supplier TCI America Supplier No. U00325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4189-02-0
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00040909
InChI Key IGBBVTAVILYDIO-MDZDMXLPSA-N
PubChem CID 5282728
ChEBI CHEBI:39450
IUPAC Name (E)-undec-2-enoic acid
SMILES CCCCCCCCC=CC(=O)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 184.28
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Undecenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.