accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCNHOHH3CNOHCOHClPdClPd(E/Z)(E/Z)

CAS RN 419581-64-9

IUPAC Name: 5-hydroxy-2-[1-(hydroxyimino)ethyl]benzen-1-ide; 6-[1-(hydroxyamino)ethylidene]-3-oxocyclohexa-1,4-dien-1-ide; bis(chloropalladiumylium)
Synonyms: 5-hydroxy-2-[1-(hydroxyimino)ethyl]benzen-1-ide; 6-[1-(hydroxyamino)ethylidene]-3-oxocyclohexa-1,4-dien-1-ide; bis(chloropalladiumylium)
Molecular Weight (g/mol): 584.05
Molecular Formula: C16H16Cl2N2O4Pd2
InChi Key: OQQFWDBYVLTJST-UHFFFAOYSA-L
SMILES: Cl[Pd+].Cl[Pd+].CC(NO)=C1C=CC(=O)C=[C-]1.CC(=NO)C1=CC=C(O)C=[C-]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results