Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) Dimer 95.0+%, TCI America™
SDP

Catalog No. D3807250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3807250MG 250 mg
D38071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3807250MG Supplier TCI America Supplier No. D3807250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 419581-64-9
Molecular Formula C16H16Cl2N2O4Pd2
Molecular Weight (g/mol) 584.054
MDL Number MFCD08705289
InChI Key OQQFWDBYVLTJST-ALDPEWTMSA-L
Synonym Najera Catalyst II
PubChem CID 134128703
IUPAC Name chloropalladium(1+);4-[1-(hydroxyamino)ethylidene]cyclohexa-2,5-dien-1-one;4-[(Z)-N-hydroxy-C-methylcarbonimidoyl]benzene-5-id-1-ol
SMILES CC(=C1C=CC(=O)C=[C-]1)NO.CC(=NO)C1=CC=C(C=[C-]1)O.Cl[Pd+].Cl[Pd+]

Specifications

Melting Point 255°C
Color Yellow
Quantity 250 mg
Formula Weight 584.05
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis[5-hydroxy-2-[1-(hydroxyimino)ethyl]phenyl]palladium(II) Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.